Assay Details
| Gene Symbol: STAT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009402.2 | NP_001009402.1 | 11 - 12 | 1605 | 66 | |
| GenBank mRNA | X78428.1 | - | 1624 | 66 | ||
More Information
Set Membership: |
|

STAT5A
Genomic Map