Assay Details
| Gene Symbol: SLIRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145184.2 | NP_001138656.1 | 1 - 2 | 106 | 74 | |
| GenBank mRNA | FJ546076.1 | - | 1 - 2 | 108 | 74 | |
More Information
Set Membership: |
|

SLIRP
Genomic Map