Assay Details
| Gene Symbol: CCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164714.1 | NP_001158186.1 | 4 - 5 | 750 | 93 | |
| GenBank mRNA | GQ268013.1 | - | 4 - 5 | 750 | 93 | |
More Information
Set Membership: |
|

CCN2
Genomic Map