Assay Details
| Gene Symbol: UGT1A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205146.2 | NP_001192075.1 | 1 - 2 | 900 | 79 | |
| GenBank mRNA | HQ263385.1 | - | 1 - 2 | 903 | 79 | |
More Information
Set Membership: |
|

UGT1A6
Genomic Map