Assay Details
| Gene Symbol: CSN1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009795.1 | NP_001009795.1 | 3 - 4 | 143 | 76 | |
| XM_012179357.3 | XP_012034747.1 | 3 - 4 | 112 | 76 | ||
| XM_027970719.1 | XP_027826520.1 | 3 - 4 | 112 | 76 | ||
| XM_027970720.1 | XP_027826521.1 | 3 - 4 | 112 | 76 | ||
| GenBank mRNA | FJ440845.1 | - | 3 - 4 | 118 | 76 | |
| FJ440846.1 | - | 3 - 4 | 118 | 76 | ||
| FJ695513.1 | - | 3 - 4 | 118 | 76 | ||
| X03237.1 | - | 143 | 76 | |||
More Information
Set Membership: |
|

CSN1S1
Genomic Map