Assay Details
| Gene Symbol: TYRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130023.1 | NP_001123495.1 | 2 - 3 | 729 | 64 | |
| GenBank mRNA | EF102109.1 | - | 2 - 3 | 708 | 64 | |
| EF102110.1 | - | 2 - 3 | 708 | 64 | ||
| EF102111.1 | - | 2 - 3 | 708 | 64 | ||
| EU760770.1 | - | 2 - 3 | 729 | 64 | ||
More Information
Set Membership: |
|

TYRP1
Genomic Map