Assay Details
| Gene Symbol: SLC26A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159756.1 | NP_001153228.1 | 1 - 2 | 671 | 75 | |
| XM_012177549.2 | XP_012032939.1 | 4 - 5 | 1613 | 75 | ||
| XM_012177551.3 | XP_012032941.1 | 4 - 5 | 1653 | 75 | ||
| XM_012177555.2 | XP_012032945.1 | 3 - 4 | 1548 | 75 | ||
More Information
Set Membership: |
|

SLC26A2
Genomic Map