Assay Details
| Gene Symbol: ZDHHC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163604.1 | NP_001157076.1 | 1 | 141 | 59 | |
| XM_027977366.1 | XP_027833167.1 | 3 | 466 | 59 | ||
| XM_027977367.1 | XP_027833168.1 | 3 | 600 | 59 | ||
| GenBank mRNA | FJ937952.1 | - | 1 | 141 | 59 | |
More Information
Set Membership: |
|

ZDHHC7
Genomic Map