Assay Details
| Gene Symbol: NUDT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202421.1 | NP_001189350.1 | 1 - 2 | 123 | 72 | |
| GenBank mRNA | HQ657210.1 | - | 1 - 2 | 123 | 72 | |
More Information
Set Membership: |
|

NUDT9
Genomic Map