Assay Details
| Gene Symbol: CITED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308562.1 | NP_001295491.1 | 2 | 925 | 76 | |
| GenBank mRNA | JX534540.1 | - | 2 | 925 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CITED2| Gene Symbol: CITED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308562.1 | NP_001295491.1 | 2 | 925 | 76 | |
| GenBank mRNA | JX534540.1 | - | 2 | 925 | 76 | |
Set Membership: |
|