Assay Details
| Gene Symbol: LOC101106534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973709.1 | XP_027829510.1 | 17 - 18 | 2351 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101106534| Gene Symbol: LOC101106534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973709.1 | XP_027829510.1 | 17 - 18 | 2351 | 87 | |
Set Membership: |
|