Assay Details
| Gene Symbol: COQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966378.1 | XP_027822179.1 | 7 - 8 | 885 | 67 | |
| XM_027966379.1 | XP_027822180.1 | 6 - 7 | 916 | 67 | ||
| XM_027966380.1 | XP_027822181.1 | 6 - 7 | 904 | 67 | ||
More Information
Set Membership: |
|

COQ4
Genomic Map