Assay Details
| Gene Symbol: CHRNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965319.1 | XP_027821120.1 | 11 - 12 | 1497 | 56 | |
| XM_027965320.1 | XP_027821121.1 | 11 - 12 | 1494 | 56 | ||
| XM_027965321.1 | XP_027821122.1 | 11 - 12 | 1392 | 56 | ||
More Information
Set Membership: |
|

CHRNG
Genomic Map