Assay Details
| Gene Symbol: LOC101108756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182071.3 | XP_012037461.2 | 5 - 6 | 1583 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101108756| Gene Symbol: LOC101108756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182071.3 | XP_012037461.2 | 5 - 6 | 1583 | 71 | |
Set Membership: |
|