Assay Details
| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015095955.2 | XP_014951441.2 | 1 - 2 | 326 | 76 | |
| XM_015095956.2 | XP_014951442.2 | 1 - 2 | 370 | 76 | ||
| XM_027969996.1 | XP_027825797.1 | 1 - 2 | 326 | 76 | ||
More Information
Set Membership: |
|

PRR16
Genomic Map