Assay Details
| Gene Symbol: FSIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010437.4 | XP_004010486.2 | 2 - 3 | 282 | 72 | |
| XM_012181177.3 | XP_012036567.2 | 2 - 3 | 282 | 72 | ||
| XM_012181178.3 | XP_012036568.2 | 2 - 3 | 282 | 72 | ||
More Information
Set Membership: |
|

FSIP1
Genomic Map