Assay Details
| Gene Symbol: SLC28A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972253.1 | XP_027828054.1 | 4 - 5 | 448 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC28A2| Gene Symbol: SLC28A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972253.1 | XP_027828054.1 | 4 - 5 | 448 | 68 | |
Set Membership: |
|