Assay Details
| Gene Symbol: LOC101105324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008102.3 | XP_004008151.2 | 2 - 3 | 460 | 88 | |
| XM_012177227.3 | XP_012032617.1 | 3 - 4 | 534 | 88 | ||
| XM_012177228.3 | XP_012032618.1 | 3 - 4 | 274 | 88 | ||
| XM_027968861.1 | XP_027824662.1 | 2 - 3 | 444 | 88 | ||
| XM_027968863.1 | XP_027824664.1 | 3 - 4 | 793 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027968863.1 transcripts.
Set Membership: |
|

LOC101105324
Genomic Map