Assay Details
| Gene Symbol: LOC101105324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008102.3 | XP_004008151.2 | 3 - 4 | 563 | 109 | |
| XM_012177227.3 | XP_012032617.1 | 4 - 5 | 637 | 109 | ||
| XM_012177228.3 | XP_012032618.1 | 4 - 5 | 377 | 109 | ||
| XM_027968861.1 | XP_027824662.1 | 3 - 4 | 547 | 109 | ||
| XM_027968862.1 | XP_027824663.1 | 2 - 3 | 170 | 109 | ||
| XM_027968863.1 | XP_027824664.1 | 4 - 5 | 896 | 109 | ||
More Information
Set Membership: |
|

LOC101105324
Genomic Map