Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969086.1 | XP_027824887.1 | 8 - 9 | 1040 | 54 | |
| XM_027969087.1 | XP_027824888.1 | 11 - 12 | 1497 | 54 | ||
| XM_027969088.1 | XP_027824889.1 | 8 - 9 | 1299 | 54 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027969086.1 transcripts.
Set Membership: |
|

DYNC2I1
Genomic Map