Assay Details
| Gene Symbol: CDK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016913.4 | XP_004016962.1 | 8 - 9 | 742 | 70 | |
| XM_027980030.1 | XP_027835831.1 | 5 - 6 | 411 | 70 | ||
| XM_027980032.1 | XP_027835833.1 | 6 - 7 | 449 | 70 | ||
More Information
Set Membership: |
|

CDK7
Genomic Map