Assay Details
| Gene Symbol: UCHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973697.1 | XP_027829498.1 | 6 - 7 | 538 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UCHL3| Gene Symbol: UCHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973697.1 | XP_027829498.1 | 6 - 7 | 538 | 62 | |
Set Membership: |
|