Assay Details
| Gene Symbol: MED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098683.2 | XP_014954169.2 | 2 - 3 | 4767 | 58 | |
| XM_027974767.1 | XP_027830568.1 | 2 - 3 | 4856 | 58 | ||
More Information
Set Membership: |
|

MED9
Genomic Map