Assay Details
| Gene Symbol: YJEFN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015095741.2 | XP_014951227.2 | 5 - 6 | 1127 | 53 | |
| XM_027969343.1 | XP_027825144.1 | 4 - 5 | 3119 | 53 | ||
More Information
Set Membership: |
|

YJEFN3
Genomic Map