Assay Details
| Gene Symbol: TBX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101661.2 | XP_014957147.2 | 7 - 8 | 2122 | 58 | |
| XM_027956552.1 | XP_027812353.1 | 6 - 7 | 2062 | 58 | ||
More Information
Set Membership: |
|

TBX3
Genomic Map