Assay Details
| Gene Symbol: LOC101102548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012142060.3 | XP_011997450.2 | 6 - 7 | 690 | 67 | |
| XM_012142071.3 | XP_011997461.2 | 6 - 7 | 690 | 67 | ||
| XM_012142079.3 | XP_011997469.2 | 6 - 7 | 690 | 67 | ||
| XM_027965072.1 | XP_027820873.1 | 6 - 7 | 690 | 67 | ||
| XM_027965074.1 | XP_027820875.1 | 6 - 7 | 690 | 67 | ||

LOC101102548
Genomic Map