Assay Details
| Gene Symbol: LOC101117364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017075.4 | XP_004017124.1 | 11 - 12 | 2440 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117364| Gene Symbol: LOC101117364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017075.4 | XP_004017124.1 | 11 - 12 | 2440 | 66 | |
Set Membership: |
|