Assay Details
| Gene Symbol: TCL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012099174.3 | XP_011954564.3 | 1 - 2 | 122 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCL1A| Gene Symbol: TCL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012099174.3 | XP_011954564.3 | 1 - 2 | 122 | 60 | |
Set Membership: |
|