Assay Details
| Gene Symbol: N4BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100171.2 | XP_014955657.2 | 3 - 4 | 2230 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

N4BP1| Gene Symbol: N4BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100171.2 | XP_014955657.2 | 3 - 4 | 2230 | 62 | |
Set Membership: |
|