Assay Details
| Gene Symbol: UBE2J1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011277.4 | XP_004011326.3 | 7 - 8 | 803 | 75 | |
| XM_012182786.3 | XP_012038176.1 | 7 - 8 | 762 | 75 | ||
More Information
Set Membership: |
|

UBE2J1
Genomic Map