Assay Details
| Gene Symbol: RAPSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979754.1 | XP_027835555.1 | 6 - 7 | 1138 | 73 | |
| XM_027979755.1 | XP_027835556.1 | 5 - 6 | 1015 | 73 | ||
| XM_027979756.1 | XP_027835557.1 | 6 - 7 | 1138 | 73 | ||
| XM_027979757.1 | XP_027835558.1 | 6 - 7 | 1138 | 73 | ||
| XM_027979758.1 | XP_027835559.1 | 5 - 6 | 799 | 73 | ||
More Information
Set Membership: |
|

RAPSN
Genomic Map