Assay Details
| Gene Symbol: DOK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094475.2 | XP_014949961.2 | 4 - 5 | 690 | 57 | |
| XM_027967026.1 | XP_027822827.1 | 4 - 5 | 510 | 57 | ||
More Information
Set Membership: |
|

DOK1
Genomic Map