Assay Details
| Gene Symbol: TMEM106A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012960.4 | XP_004013009.2 | 5 - 6 | 688 | 59 | |
| XM_012186167.3 | XP_012041557.2 | 5 - 6 | 952 | 59 | ||
More Information
Set Membership: |
|

TMEM106A
Genomic Map