Assay Details
| Gene Symbol: ZNF532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960889.1 | XP_027816690.1 | 9 - 10 | 3916 | 59 | |
| XM_027960890.1 | XP_027816691.1 | 10 - 11 | 3878 | 59 | ||
| XM_027960891.1 | XP_027816692.1 | 9 - 10 | 3899 | 59 | ||
| XM_027960892.1 | XP_027816693.1 | 10 - 11 | 3708 | 59 | ||
| XM_027960893.1 | XP_027816694.1 | 10 - 11 | 3712 | 59 | ||
| XM_027960894.1 | XP_027816695.1 | 10 - 11 | 3791 | 59 | ||
| XM_027960895.1 | XP_027816696.1 | 10 - 11 | 4650 | 59 | ||
More Information
Set Membership: |
|

ZNF532
Genomic Map