Assay Details
| Gene Symbol: DDX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002323.4 | XP_004002372.2 | 8 - 9 | 1284 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDX20| Gene Symbol: DDX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002323.4 | XP_004002372.2 | 8 - 9 | 1284 | 66 | |
Set Membership: |
|