Assay Details
| Gene Symbol: TXNL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020395.3 | XP_004020444.1 | 2 - 3 | 298 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TXNL4A| Gene Symbol: TXNL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020395.3 | XP_004020444.1 | 2 - 3 | 298 | 93 | |
Set Membership: |
|