Assay Details
| Gene Symbol: LOC114116147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972595.1 | XP_027828396.1 | 2 - 3 | 242 | 65 | |
| Gene Symbol: C8H6orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972594.1 | XP_027828395.1 | 6 - 7 | 605 | 65 | |

LOC114116147
Genomic Map