Assay Details
| Gene Symbol: LOC101121099 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972246.1 | XP_027828047.1 | 3 - 4 | 406 | 79 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101121099| Gene Symbol: LOC101121099 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972246.1 | XP_027828047.1 | 3 - 4 | 406 | 79 | |