Assay Details
| Gene Symbol: SLC13A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012497.4 | XP_004012546.2 | 11 - 12 | 1803 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC13A2| Gene Symbol: SLC13A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012497.4 | XP_004012546.2 | 11 - 12 | 1803 | 61 | |
Set Membership: |
|