Assay Details
| Gene Symbol: MRPL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974097.1 | XP_027829898.1 | 8 - 9 | 1046 | 66 | |
| XM_027974098.1 | XP_027829899.1 | 8 - 9 | 1089 | 66 | ||
More Information
Set Membership: |
|

MRPL38
Genomic Map