Assay Details
| Gene Symbol: HACD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964572.1 | XP_027820373.1 | 5 - 6 | 610 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HACD4| Gene Symbol: HACD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964572.1 | XP_027820373.1 | 5 - 6 | 610 | 102 | |
Set Membership: |
|