Assay Details
| Gene Symbol: POLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013031.3 | XP_004013080.2 | 2 - 3 | 937 | 75 | |
| XM_012186340.2 | XP_012041730.2 | 2 - 3 | 937 | 75 | ||
More Information
Set Membership: |
|

POLG2
Genomic Map