Assay Details
| Gene Symbol: LOC101103973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177031.3 | XP_012032421.2 | 16 - 17 | 1878 | 70 | |
| XM_027968763.1 | XP_027824564.1 | 15 - 16 | 4477 | 70 | ||
| XM_027968764.1 | XP_027824565.1 | 13 - 14 | 1535 | 70 | ||
| XM_027968765.1 | XP_027824566.1 | 14 - 15 | 1464 | 70 | ||
More Information
Set Membership: |
|

LOC101103973
Genomic Map