Assay Details
| Gene Symbol: GRAMD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008666.4 | XP_004008715.2 | 12 - 13 | 1268 | 68 | |
| XM_012178205.3 | XP_012033595.2 | 12 - 13 | 1304 | 68 | ||
More Information
Set Membership: |
|

GRAMD2B
Genomic Map