Assay Details
| Gene Symbol: LOC114117592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976874.1 | XP_027832675.1 | 2 - 3 | 522 | 68 | |
| XM_027976875.1 | XP_027832676.1 | 2 - 3 | 524 | 68 | ||
| XM_027976876.1 | XP_027832677.1 | 2 - 3 | 524 | 68 | ||
| Gene Symbol: LOC101103001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976879.1 | XP_027832680.1 | 2 - 3 | 521 | 68 | |

LOC114117592
Genomic Map