Assay Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020947.4 | XP_004020996.1 | 5 - 6 | 927 | 66 | |
| XM_015104194.2 | XP_014959680.1 | 4 - 5 | 950 | 66 | ||
| XM_027961846.1 | XP_027817647.1 | 3 - 4 | 4305 | 66 | ||
More Information
Set Membership: |
|

TMEM248
Genomic Map