Assay Details
| Gene Symbol: PHYHIPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021385.4 | XP_004021434.1 | 3 - 4 | 832 | 95 | |
| XM_012105098.3 | XP_011960488.1 | 3 - 4 | 522 | 95 | ||
| XM_027962265.1 | XP_027818066.1 | 3 - 4 | 632 | 95 | ||
More Information
Set Membership: |
|

PHYHIPL
Genomic Map