Assay Details
| Gene Symbol: AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019719.4 | XP_004019768.1 | 2 - 3 | 413 | 59 | |
| XM_012102185.3 | XP_011957575.2 | 2 - 3 | 413 | 59 | ||
More Information
Set Membership: |
|

AIP
Genomic Map