Assay Details
| Gene Symbol: MBTPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963340.1 | XP_027819141.1 | 2 - 3 | 320 | 62 | |
| XM_027963341.1 | XP_027819142.1 | 2 - 3 | 323 | 62 | ||
More Information
Set Membership: |
|

MBTPS2
Genomic Map