Assay Details
| Gene Symbol: MBTPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963340.1 | XP_027819141.1 | 4 - 5 | 633 | 83 | |
| XM_027963341.1 | XP_027819142.1 | 4 - 5 | 636 | 83 | ||
| Gene Symbol: YY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963342.1 | XP_027819143.1 | 1 - 2 | 2730 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027963342.1 transcripts.
Set Membership: |
|

MBTPS2
Genomic Map